Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01185283_m1
          See other CNKSR1 GE Assays ›
          Gene Symbol:
          Cnksr1
          Gene Name:
          connector enhancer of kinase suppressor of Ras 1
          Gene Aliases:
          BC037758
          Chromosome Location:
          Chr.4: 134228041 - 134238416 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01185283_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cnksr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081047.1 NP_001074516.1 14 - 15 1375 100
          GenBank mRNA AK145910.1 - 14 - 15 1337 100
          BC113805.1 - 14 - 15 1340 40062208 100
          BC113956.1 - 14 - 15 1340 40062209 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003731.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          Ras protein signal transduction
          Rho protein signal transduction
          protein binding, bridging

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P140974 BC113805.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P140974 BC113956.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P140974 NM_001081047.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P140974 AK145910.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P140974 BC037758.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 289031 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 154628 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764530 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 20896 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 256613 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 562304 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143820 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101878 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P140974 BC113805.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P140974 BC113956.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P140974 NM_001081047.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P140974 AK145910.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P140974 BC037758.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05411371 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101878 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101878 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03884441 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03884442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03884462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03247042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03280705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03739669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03598321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03884456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03280706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03598322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03884466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03456864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03889250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03840367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03548765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321708 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10517220 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517231 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517217 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517235 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517219 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517215 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517223 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517230 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517228 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517222 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517226 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517224 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517229 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517225 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517234 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517221 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517214 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517227 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517232 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517218 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517216 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517233 - -
          Affymetrix HT MG-430 PM Array Plate 1455399_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10517213 - -
          Mouse Genome 430 2.0 Array 1455399_at - -
          Mouse Genome 430 2.0 Array 1455399_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 194231_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 194231_at - -
          Affymetrix Mouse Clariom S Array PSR0400030692.mm.1 TC0400003731.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030692.mm.1 NM_001081047.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030692.mm.1 NM_001081047.1 Refseq
          Mouse Expression Set 430 1455399_at - -
          Mouse Expression Set 430 1455399_PM_at - -
          Murine Genome U74v2 Set 129176_PM_at - -
          Murine Genome U74v2 Set 129176_at - -
          Affymetrix HT Mouse Genome 430B Array 1455399_at - -
          Affymetrix HT Mouse Genome 430B Array 1455399_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2723329 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2723329 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0