Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Aim1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162970.1 | NP_001156442.1 | 15 - 16 | 3990 | 79 | |
GenBank mRNA | AK145690.1 | - | 15 - 16 | 3996 | 79 | |
AK155034.1 | - | 3 - 4 | 341 | 79 | ||
AK165037.1 | - | 2 - 3 | 265 | 79 | ||
BC042709.1 | - | 10 - 11 | 1011 | 79 | ||
BC150738.1 | - | 14 - 15 | 3919 | 9087649 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |