| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23254145 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23254146 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23254147 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23254151 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23254148 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2034385 |
NM_001005788.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2034385 |
BC066020.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2034385 |
U29497.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
671973 |
AB067528.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
671973 |
AF459815.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
671973 |
AF459816.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
671973 |
AK028986.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
671973 |
AK078952.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
671973 |
NM_033541.4 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
529866 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
685658 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
650410 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
144914 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
558703 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
715072 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
833064 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
117143 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
692698 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
590965 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
132836 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
142685 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P144479 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P118439 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2034385 |
NM_001005788.3 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2034385 |
BC066020.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2034385 |
U29497.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254144 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254162 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254125 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254160 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254158 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254152 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254133 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254130 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254119 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254153 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254134 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254135 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254165 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254148 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254150 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254120 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254156 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254138 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254155 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254139 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254137 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254145 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254151 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254136 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254072 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254140 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254142 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254157 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254146 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254143 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254127 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254167 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254161 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05380188 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P144479 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00973909 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P118439 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P478550 |
NM_001005788.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P478550 |
BC066020.1 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P144479 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P118439 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03782836 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03308044 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03308042 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03308041 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03782835 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03782834 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23254145 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23254147 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23254148 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23254146 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23254151 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254027 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254077 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254160 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254096 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254148 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254038 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254074 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254181 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254152 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254157 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254093 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254100 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254047 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254138 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254043 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254034 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254059 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254099 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254031 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254134 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254081 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254135 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254103 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254014 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254052 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254087 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254171 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254085 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254140 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254018 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254115 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254145 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254011 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254067 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254016 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254101 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254078 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254105 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254010 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254030 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254139 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254177 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254076 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254153 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254025 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254102 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254183 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254126 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254057 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254169 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254146 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254133 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254130 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254032 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254094 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254123 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254161 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254055 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254041 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254110 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254037 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254142 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254185 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254035 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254046 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254045 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254019 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254184 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254144 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254155 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254024 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254033 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254091 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254086 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254176 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254080 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254083 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254151 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254182 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254050 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254072 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254126 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03928091 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03824244 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03674336 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254163 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03917903 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03782835 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03783523 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03824246 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254106 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254085 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254114 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03361414 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254092 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03928085 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254115 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254140 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254130 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03490829 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03741034 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254167 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03917906 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254187 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254089 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254133 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254157 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254095 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254074 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254145 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254118 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254158 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03780471 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03308041 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04018525 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254097 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03824248 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254169 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254103 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254117 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254168 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254101 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254099 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254120 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03719305 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254081 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254125 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254138 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03428920 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03783524 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254155 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03534908 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03782834 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254112 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03381710 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03824251 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254096 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254172 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03534907 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254153 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254080 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254091 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03626845 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254142 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254173 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254078 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03694292 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03741035 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254075 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254087 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03308044 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03428921 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254098 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254071 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03917907 |
- |
- |