| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400029682.mm.1 |
TC0400003628.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400029682.mm.1 |
NM_001034872.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400029682.mm.1 |
NM_001034872.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400029680.mm.1 |
TC0400003628.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400029680.mm.1 |
NM_001034872.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400029680.mm.1 |
NM_001034872.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400029681.mm.1 |
TC0400003628.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400029681.mm.1 |
NM_001034872.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400029681.mm.1 |
NM_001034872.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400029684.mm.1 |
TC0400003628.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400029684.mm.1 |
NM_001034872.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400029684.mm.1 |
NM_001034872.2 |
Refseq |