Search
Search

Hcrtr1| Gene Symbol: Hcrtr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163027.1 | NP_001156499.1 | 6 - 7 | 1028 | 66 | |
| NM_198959.2 | NP_945197.2 | 6 - 7 | 1054 | 66 | ||
| GenBank mRNA | AF394596.1 | - | 2 - 3 | 344 | 66 | |
| AK140137.1 | - | 6 - 7 | 1028 | 66 | ||
| AY255599.1 | - | 2 - 3 | 322 | 66 | ||
| AY336083.1 | - | 6 - 7 | 1054 | 66 | ||
| BC119582.2 | - | 6 - 7 | 926 | 66 | ||
| BC119583.2 | - | 5 - 6 | 910 | 40058834 | 66 | |
Set Membership: |
|