Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01186750_m1
          See other BC064078 GE Assays ›
          Gene Symbol:
          BC064078
          Gene Name:
          cDNA sequence BC064078
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 128993020 - 129006647 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01186750_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BC064078
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015455.1 2 - 3 204 102
          GenBank mRNA BC016205.1 - 2 - 3 217 102
          BC017132.1 - 2 - 3 217 102
          BC025095.1 - 2 - 3 201 102
          BC038635.1 - 2 - 3 200 102
          BC051485.1 - 2 - 3 263 102
          BC064078.1 - 2 - 3 193 102
          BC096405.1 - 2 - 3 204 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001524.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2154719 AK134020.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2154719 AK134020.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01445067 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01445062 - -
          Whole Mouse Genome G4122A A_52_P812362 BC051485.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P812362 BC064078.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P812362 BC096405.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P812362 NR_015455.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P812362 AK134020.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24841567 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1444632_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1435069_PM_at - -
          Mouse Genome 430 2.0 Array 1444632_at - -
          Mouse Genome 430 2.0 Array 1444632_PM_at - -
          Mouse Genome 430 2.0 Array 1435069_at - -
          Mouse Genome 430 2.0 Array 1435069_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 408064_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 408064_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600012413.mm.1 TC0600001524.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012413.mm.1 NR_015455.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012415.mm.1 TC0600001524.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012415.mm.1 NR_015455.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012418.mm.1 TC0600001524.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012418.mm.1 NR_015455.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012419.mm.1 TC0600001524.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012419.mm.1 NR_015455.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012420.mm.1 TC0600001524.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012420.mm.1 NR_015455.2 Refseq
          Mouse Expression Set 430 1444632_at - -
          Mouse Expression Set 430 1435069_at - -
          Mouse Expression Set 430 1444632_PM_at - -
          Mouse Expression Set 430 1435069_PM_at - -
          Murine Genome U74v2 Set 132397_at - -
          Murine Genome U74v2 Set 132397_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435069_at - -
          Affymetrix HT Mouse Genome 430B Array 1444632_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444632_at - -
          Affymetrix HT Mouse Genome 430B Array 1435069_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0