Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01187363_m1
          See other XKR8 GE Assays ›
          Gene Symbol:
          Xkr8
          Gene Name:
          X Kell blood group precursor related family member 8 homolog
          Gene Aliases:
          4931440N07Rik, AI597014, Gm1031, XRG8, mXkr8
          Chromosome Location:
          Chr.4: 132724904 - 132732614 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01187363_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Xkr8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_201368.1 NP_958756.1 2 - 3 566 81
          GenBank mRNA AK029920.1 - 2 - 3 566 81
          AY534254.1 - 2 - 3 566 81
          BC057035.1 - 2 - 3 502 5698137 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003679.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Xkr8

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          engulfment of apoptotic cell
          phosphatidylserine exposure on apoptotic cell surface
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053032 AY534254.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053032 BC057035.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053032 NM_201368.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053032 AK029920.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297940 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 685810 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 682017 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 561396 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 959170 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108994 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113228 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156920 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2053032 AY534254.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2053032 BC057035.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2053032 NM_201368.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2053032 AK029920.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23313778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23313763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23313774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23313772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23313773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23313768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23313770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23313779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23313777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23313780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23313764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23313776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23313762 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108994 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156920 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113228 - -
          Whole Mouse Genome G4122A A_51_P389705 BC057035.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P389705 NM_201368.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P389705 AK029920.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123675 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156920 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108994 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113228 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03887289 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03887300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23313747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03484336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03887290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03484316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03330128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03216377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313751 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03887294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03312952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03887300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03685163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03887298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23313763 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10516954 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516953 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516951 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516952 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516955 - -
          Affymetrix HT MG-430 PM Array Plate 1455704_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1439590_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10516950 - -
          Mouse Genome 430 2.0 Array 1455704_PM_at - -
          Mouse Genome 430 2.0 Array 1439590_PM_at - -
          Mouse Genome 430 2.0 Array 1455704_at - -
          Mouse Genome 430 2.0 Array 1439590_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 381560_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 381560_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400030164.mm.1 TC0400003679.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030164.mm.1 NM_201368.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030164.mm.1 NM_201368.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030165.mm.1 TC0400003679.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030165.mm.1 NM_201368.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030165.mm.1 NM_201368.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030162.mm.1 TC0400003679.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030162.mm.1 NM_201368.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030162.mm.1 NM_201368.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030167.mm.1 TC0400003679.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030167.mm.1 NM_201368.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030167.mm.1 NM_201368.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030168.mm.1 TC0400003679.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030168.mm.1 NM_201368.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030168.mm.1 NM_201368.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030170.mm.1 TC0400003679.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030170.mm.1 NM_201368.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030170.mm.1 NM_201368.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030163.mm.1 TC0400003679.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030163.mm.1 NM_201368.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030163.mm.1 NM_201368.1 Refseq
          Mouse Expression Set 430 1455704_at - -
          Mouse Expression Set 430 1439590_PM_at - -
          Mouse Expression Set 430 1439590_at - -
          Mouse Expression Set 430 1455704_PM_at - -
          Murine Genome U74v2 Set 138121_at - -
          Murine Genome U74v2 Set 138121_PM_at - -
          Murine Genome U74v2 Set 170874_PM_i_at - -
          Murine Genome U74v2 Set 170874_i_at - -
          Murine Genome U74v2 Set 132110_PM_s_at - -
          Murine Genome U74v2 Set 132110_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1439590_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439590_at - -
          Affymetrix HT Mouse Genome 430B Array 1455704_at - -
          Affymetrix HT Mouse Genome 430B Array 1455704_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 700718 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1190575 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001190575 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1254917 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254917 - -
          mouse-6 v1.1 (Illumina) scl0381560.1_274-S - -
          Illumina Sentrix mouse-6.v1 bead array 1190575 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0381560.1_274-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1190575 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2662990 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2662990 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline