Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01187764_m1
          See other AP1B1 GE Assays ›
          Gene Symbol:
          Ap1b1
          Gene Name:
          adaptor protein complex AP-1, beta 1 subunit
          Gene Aliases:
          Adtb1, b2b1660Clo
          Chromosome Location:
          Chr.11: 4947521 - 5042794 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01187764_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ap1b1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001243043.1 NP_001229972.1 8 - 9 1511 112
          NM_001243044.1 NP_001229973.1 8 - 9 1314 112
          NM_007454.3 NP_031480.2 8 - 9 1314 112
          GenBank mRNA AK034140.1 - 8 - 9 1514 112
          AK155892.1 - 8 - 9 1234 112
          AK159273.1 - 8 - 9 1205 112
          AK160042.1 - 8 - 9 1205 112
          AK165747.1 - 8 - 9 1231 112
          AK170376.1 - 8 - 9 1225 112
          BC008513.1 - 8 - 9 1127 3601435 112
          CA315828.1 - 3 - 4 534 112
          Y07919.1 - 8 - 9 1169 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000058.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ap1b1

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          intracellular protein transport
          protein transport
          vesicle-mediated transport
          clathrin coat assembly
          protein binding
          protein transporter activity
          protein kinase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100000668.mm.1 NM_001243043.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000668.mm.1 NM_001243043.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000668.mm.1 NM_001243044.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000668.mm.1 NM_001243044.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000668.mm.1 NM_007454.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000668.mm.1 NM_007454.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000668.mm.1 TC1100000058.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000669.mm.1 NM_001243043.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000669.mm.1 NM_001243043.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000669.mm.1 NM_001243044.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000669.mm.1 NM_001243044.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000669.mm.1 NM_007454.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000669.mm.1 NM_007454.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000669.mm.1 TC1100000058.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0