Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01187768_m1
          See other PGAM2 GE Assays ›
          Gene Symbol:
          Pgam2
          Gene Name:
          phosphoglycerate mutase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 5801637 - 5803796 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01187768_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pgam2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_018870.3 NP_061358.1 2 - 3 710 84
          GenBank mRNA AF029843.1 - 2 - 3 647 84
          AK146703.1 - 2 - 3 637 84
          AK169019.1 - 2 - 3 637 84
          BC010750.1 - 2 - 3 615 4235174 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002152.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pgam2

          Panther Classification:

          Molecular Function -

          mutase

          Gene Ontology Categories:

          Function(s) Process(es)

          gluconeogenesis
          glycolytic process
          striated muscle contraction
          Notch signaling pathway
          metabolic process
          response to inorganic substance
          response to mercury ion
          catalytic activity
          bisphosphoglycerate mutase activity
          bisphosphoglycerate 2-phosphatase activity
          phosphoglycerate mutase activity
          protein binding
          hydrolase activity
          isomerase activity
          intramolecular transferase activity, phosphotransferases
          2,3-bisphosphoglycerate-dependent phosphoglycerate mutase activity
          cofactor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P264495 AF029843.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P264495 AK146703.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P264495 AK169019.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P264495 NM_018870.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P264495 AV041299.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P264495 AF029843.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P264495 AK146703.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P264495 AK169019.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P264495 NM_018870.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P264495 AV041299.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 821441 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158152 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P264495 AF029843.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P264495 AK146703.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P264495 AK169019.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P264495 NM_018870.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P264495 AV041299.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P264495 AF029843.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P264495 AK146703.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P264495 AK169019.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P264495 NM_018870.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P264495 AV041299.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P264495 AF029843.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P264495 AK146703.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P264495 AK169019.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P264495 NM_018870.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P264495 AV041299.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P264495 AF029843.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P264495 AK146703.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P264495 AK169019.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P264495 NM_018870.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P264495 AV041299.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27565056 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158152 - -
          Whole Mouse Genome G4122A A_51_P264495 AF029843.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264495 AK146703.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264495 AK169019.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264495 NM_018870.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264495 AV041299.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264495 AF029843.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264495 AK146703.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264495 AK169019.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264495 NM_018870.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264495 AV041299.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109808 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158152 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09273094 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09273098 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09273093 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09273059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27564994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27564977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27564984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27564990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27564981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27564987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27564998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27564982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27564992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27564979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27564991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27564985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27565072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09273087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09273081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09273084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09273073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09273086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27565071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09273060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09538833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08975430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09733133 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10384017 - -
          Affymetrix Mouse Gene 1.0 ST Array 10384019 - -
          Affymetrix Mouse Gene 1.0 ST Array 10384016 - -
          Affymetrix Mouse Gene 1.0 ST Array 10384018 - -
          Affymetrix HT MG-430 PM Array Plate 1418373_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10384015 - -
          Mouse Genome 430 2.0 Array 1418373_at - -
          Mouse Genome 430 2.0 Array 1418373_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56012_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56012_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100020902.mm.1 NM_018870.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020902.mm.1 NM_018870.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020902.mm.1 TC1100002152.mm.1 MTA TC
          Mouse Expression Set 430 1418373_at - -
          Mouse Expression Set 430 1418373_PM_at - -
          Murine Genome U74v2 Set 92599_PM_at - -
          Murine Genome U74v2 Set 92599_at - -
          Mouse Genome 430A 2.0 Array 1418373_at - -
          Mouse Genome 430A 2.0 Array 1418373_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418373_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418373_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 733947 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3610605 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003610605 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2588815 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2588815 - -
          mouse-6 v1.1 (Illumina) scl40555.3.1_120-S - -
          Illumina Sentrix mouse-6.v1 bead array 3610605 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40555.3.1_120-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3610605 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2588815 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2588815 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl40555.3.1_120-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline