Search
Search

2210015D19Rik| Gene Symbol: 2210015D19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015577.1 | 1 - 2 | 90 | 83 | ||
| GenBank mRNA | AK005556.1 | - | 1 - 2 | 90 | 83 | |
| AK008730.1 | - | 1 - 2 | 85 | 83 | ||
| AK009198.1 | - | 1 - 2 | 112 | 83 | ||
| BC052911.1 | - | 1 - 2 | 74 | 83 | ||
| BC094935.1 | - | 1 - 2 | 62 | 83 | ||
Set Membership: |
|