Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zpbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185153.1 | NP_001172082.1 | 3 - 4 | 387 | 63 | |
NM_015785.2 | NP_056600.1 | 3 - 4 | 558 | 63 | ||
GenBank mRNA | BC139450.1 | - | 3 - 4 | 527 | 8862472 | 63 |
BC139453.1 | - | 3 - 4 | 527 | 8862475 | 63 | |
BC145628.1 | - | 3 - 4 | 356 | 9054196 | 63 | |
D17569.1 | - | 3 - 4 | 547 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |