Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01187858_m1
          See other EGFR GE Assays ›
          Gene Symbol:
          Egfr
          Gene Name:
          epidermal growth factor receptor
          Gene Aliases:
          9030024J15Rik, AI552599, Erbb, Errb1, Errp, Wa5, wa-2, wa2
          Chromosome Location:
          Chr.11: 16752203 - 16913907 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01187858_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Egfr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007912.4 NP_031938.1 2 - 3 521 101
          NM_207655.2 NP_997538.1 2 - 3 521 101
          GenBank mRNA AF124513.1 - 2 - 3 455 101
          AF275367.1 - 2 - 3 464 101
          AK004883.1 - 2 - 3 522 101
          AK004911.1 - 2 - 3 496 101
          AK004944.1 - 2 - 3 515 101
          AK134262.1 - 2 - 3 495 101
          BC023729.1 - 2 - 3 492 5351807 101
          L06864.1 - 2 - 3 207 101
          U03425.1 - 2 - 3 291 101
          X59698.1 - 2 - 3 400 101
          X78987.1 - 2 - 3 300 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000161.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Egfr

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPKK activity
          cell morphogenesis
          embryonic placenta development
          positive regulation of protein phosphorylation
          hair follicle development
          translation
          protein phosphorylation
          signal transduction
          cell surface receptor signaling pathway
          transmembrane receptor protein tyrosine kinase signaling pathway
          epidermal growth factor receptor signaling pathway
          multicellular organism development
          salivary gland morphogenesis
          learning or memory
          cell proliferation
          positive regulation of cell proliferation
          epidermis development
          magnesium ion homeostasis
          phosphorylation
          single organismal cell-cell adhesion
          peptidyl-tyrosine phosphorylation
          cerebral cortex cell migration
          positive regulation of cell migration
          positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
          positive regulation of superoxide anion generation
          positive regulation of catenin import into nucleus
          intracellular signal transduction
          regulation of cell proliferation
          negative regulation of protein catabolic process
          positive regulation of phosphorylation
          ovulation cycle
          negative regulation of apoptotic process
          positive regulation of MAP kinase activity
          positive regulation of nitric oxide biosynthetic process
          positive regulation of DNA repair
          positive regulation of DNA replication
          positive regulation of vasoconstriction
          positive regulation of vasodilation
          negative regulation of mitotic cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          protein autophosphorylation
          astrocyte activation
          positive regulation of fibroblast proliferation
          digestive tract morphogenesis
          positive regulation of smooth muscle cell proliferation
          neuron projection morphogenesis
          positive regulation of epithelial cell proliferation
          positive regulation of inflammatory response
          regulation of peptidyl-tyrosine phosphorylation
          regulation of nitric-oxide synthase activity
          response to calcium ion
          positive regulation of protein kinase B signaling
          positive regulation of synaptic transmission, glutamatergic
          morphogenesis of an epithelial fold
          response to UV-A
          positive regulation of ERK1 and ERK2 cascade
          cellular response to amino acid stimulus
          cellular response to epidermal growth factor stimulus
          cellular response to estradiol stimulus
          nucleotide binding
          glycoprotein binding
          chromatin binding
          protein kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          receptor signaling protein tyrosine kinase activity
          signal transducer activity
          transmembrane signaling receptor activity
          epidermal growth factor-activated receptor activity
          receptor binding
          integrin binding
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          enzyme binding
          protein kinase binding
          protein phosphatase binding
          nitric-oxide synthase regulator activity
          ubiquitin protein ligase binding
          identical protein binding
          protein heterodimerization activity
          epidermal growth factor binding
          actin filament binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100001438.mm.1 NM_007912.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100001438.mm.1 NM_007912.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100001438.mm.1 NM_207655.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100001438.mm.1 NM_207655.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100001438.mm.1 TC1100000161.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline