Search
Search

Zfp362| Gene Symbol: Zfp362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081098.1 | NP_001074567.1 | 4 - 5 | 539 | 154 | |
| GenBank mRNA | BC147222.1 | - | 3 - 4 | 438 | 9056279 | 154 |
| BC147223.1 | - | 3 - 4 | 438 | 9056281 | 154 | |
| BC150964.1 | - | 3 - 4 | 438 | 9087875 | 154 | |
Set Membership: |
|