Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01188000_m1
          See other RNF19B GE Assays ›
          Gene Symbol:
          Rnf19b
          Gene Name:
          ring finger protein 19B
          Gene Aliases:
          4930534K13Rik, 4930555L03Rik, Ibrdc3
          Chromosome Location:
          Chr.4: 129058174 - 129085886 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01188000_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf19b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029219.1 NP_083495.1 8 - 9 1735 76
          GenBank mRNA AK015966.1 - 3 - 4 419 76
          BC052529.1 - 2 - 3 337 76
          BC117807.1 - 8 - 9 1293 76
          BC139808.1 - 8 - 9 1296 76

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          natural killer cell mediated cytotoxicity
          interferon-gamma secretion
          zinc ion binding
          UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
          ligase activity
          ribosomal S6-glutamic acid ligase activity
          coenzyme F420-0 gamma-glutamyl ligase activity
          coenzyme F420-2 alpha-glutamyl ligase activity
          metal ion binding
          protein-glycine ligase activity
          protein-glycine ligase activity, initiating
          protein-glycine ligase activity, elongating
          tubulin-glycine ligase activity
          protein-glutamic acid ligase activity
          tubulin-glutamic acid ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P124724 BC117807.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P124724 BC139808.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P124724 AK016134.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2068607 BC052529.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2068607 NM_029219.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 25271 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 364683 AK044354.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 397876 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309185 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 219735 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 428407 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 955285 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 95787 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 406408 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679879 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 207005 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 263086 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418292 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 434351 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 195777 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701076 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 677146 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 29320 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171212 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147630 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127119 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2068607 BC052529.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2068607 NM_029219.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P124724 BC117807.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P124724 BC139808.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P124724 AK016134.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171212 - -
          Whole Mouse Genome G4122A A_51_P475155 AK015966.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P475156 BC139808.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P475156 AK015966.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03735970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03868125 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03530789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03695079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03269595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294821 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03530793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03850757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03269612 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03430344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03695085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03850759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03695078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03972313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03239954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03868124 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03239957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03269599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03269596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03530790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03530792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03530784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03239964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03296896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03868122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03269613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294823 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10508403 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508397 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508394 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508398 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508401 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508393 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508402 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508404 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508395 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508399 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508396 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508400 - -
          Affymetrix HT MG-430 PM Array Plate 1432478_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1435226_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10508392 - -
          Mouse Genome 430 2.0 Array 1435226_PM_at - -
          Mouse Genome 430 2.0 Array 1432478_PM_a_at - -
          Mouse Genome 430 2.0 Array 1435226_at - -
          Mouse Genome 430 2.0 Array 1432478_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75234_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75234_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400011630.mm.1 NM_029219.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011630.mm.1 NM_029219.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011630.mm.1 TC0400001510.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011632.mm.1 NM_029219.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011632.mm.1 NM_029219.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011632.mm.1 TC0400001510.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011629.mm.1 NM_029219.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011629.mm.1 NM_029219.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011629.mm.1 TC0400001510.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011631.mm.1 NM_029219.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011631.mm.1 NM_029219.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011631.mm.1 TC0400001510.mm.1 MTA TC
          Mouse Expression Set 430 1435226_PM_at - -
          Mouse Expression Set 430 1432478_PM_a_at - -
          Mouse Expression Set 430 1435226_at - -
          Mouse Expression Set 430 1432478_a_at - -
          Murine Genome U74v2 Set 103563_PM_at - -
          Murine Genome U74v2 Set 103563_at - -
          Mouse Genome 430A 2.0 Array 1432478_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1435226_at - -
          Mouse Genome 430A 2.0 Array 1432478_a_at - -
          Mouse Genome 430A 2.0 Array 1435226_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1435226_at - -
          Affymetrix HT Mouse Genome 430A Array 1435226_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1432478_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1432478_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 695308 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2700689 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2700689 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline