Search
Search

Lypla2| Gene Symbol: Lypla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011942.1 | NP_036072.1 | 5 - 6 | 326 | 61 | |
| GenBank mRNA | AB009653.1 | - | 5 - 6 | 326 | 61 | |
| AK003689.1 | - | 5 - 6 | 293 | 61 | ||
| AK075590.1 | - | 5 - 6 | 318 | 61 | ||
| AK089112.1 | - | 5 - 6 | 318 | 61 | ||
| AK167478.1 | - | 5 - 6 | 313 | 61 | ||
| BC068120.1 | - | 5 - 6 | 352 | 5714650 | 61 | |
Set Membership: |
|