Search
Search

C77080| Gene Symbol: C77080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033189.3 | NP_001028361.1 | 2 - 3 | 493 | 79 | |
| NM_001285865.1 | NP_001272794.1 | 2 - 3 | 383 | 79 | ||
| NM_001285866.1 | NP_001272795.1 | 2 - 3 | 355 | 79 | ||
| NM_001285867.1 | NP_001272796.1 | 2 - 3 | 244 | 79 | ||
| GenBank mRNA | AK038020.1 | - | 2 - 3 | 334 | 79 | |
| AK129382.1 | - | 2 - 3 | 331 | 79 | ||
| AK154456.1 | - | 2 - 3 | 245 | 79 | ||
| AK154932.1 | - | 2 - 3 | 494 | 79 | ||
| BY201655.1 | - | 2 - 3 | 242 | 79 | ||
| CJ106414.1 | - | 2 - 3 | 384 | 79 | ||
Set Membership: |
|