Search
Search
| Gene Symbol: Snca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042451.2 | NP_001035916.1 | 3 - 4 | 471 | 67 | |
| NM_009221.2 | NP_033247.1 | 3 - 4 | 375 | 67 | ||
| GenBank mRNA | AF033261.1 | - | 3 - 4 | 298 | 67 | |
| AF179273.1 | - | 3 - 4 | 375 | 67 | ||
| AK156316.1 | - | 3 - 4 | 307 | 67 | ||
| AK205246.1 | - | 2 - 3 | 197 | 67 | ||
| BC046764.1 | - | 3 - 4 | 214 | 5693785 | 67 | |
Set Membership: |
|