Search
Search

0610030E20Rik| Gene Symbol: 0610030E20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026696.1 | NP_080972.1 | 2 - 3 | 317 | 75 | |
| GenBank mRNA | AK002701.1 | - | 2 - 3 | 263 | 75 | |
| BB844069.1 | - | 2 - 3 | 317 | 75 | ||
| BC064080.1 | - | 2 - 3 | 300 | 1246433 | 75 | |
| BC066053.1 | - | 2 - 3 | 291 | 6827710 | 75 | |
| BC117811.1 | - | 1 - 2 | 344 | 75 | ||
| BC117812.1 | - | 2 - 3 | 267 | 40092656 | 75 | |
Set Membership: |
|