Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01189458_g1
          See other IGKV2-112 GE Assays ›
          Gene Symbol:
          Igkv2-112
          Gene Name:
          immunoglobulin kappa variable 2-112
          Gene Aliases:
          EG381776, Gm5153, hg24
          Chromosome Location:
          Chr.6: 68219981 - 68220705 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01189458_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Igkv2-112
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA M19909.1 - 1 - 2 93 88
          M19910.1 - 1 - 2 81 88

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          immunoglobulin

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P559140 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24524250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24524268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24524291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05120095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05224485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24524265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24524262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05224468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24524288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05224477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24524298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24524284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24524256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24524277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05015559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24524283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05120101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05224464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24524292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05015557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05224489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24524279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05224491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24524287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24524281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05822475 - -
          Array Name Probe IDs Targets Target Type
          Murine Genome U74v2 Set 96969_at - -
          Murine Genome U74v2 Set 96969_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 763433 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252889 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline