Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2159969 |
NM_001104642.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
720507 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
647027 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
411003 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
631193 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
353951 |
AK035990.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
353951 |
NM_177597.4 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
769874 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572960 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
933650 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
49939 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
528934 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
124640 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
367341 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
806515 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
517251 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
66768 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
51309 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
16172 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
179279 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2159969 |
NM_001104642.1 |
mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814348 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814296 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814351 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814261 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814211 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814219 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814323 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814298 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814347 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814315 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814222 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814206 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814337 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814253 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814271 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814345 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814224 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814293 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814210 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814260 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814336 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814309 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814258 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814287 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814302 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814297 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814257 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814335 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814294 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814274 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814231 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814346 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24814242 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24814337 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05520281 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24814348 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05219156 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05819814 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24814327 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24814353 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24814328 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05118922 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05674929 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05638982 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05520283 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05819816 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24814332 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24814316 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24814351 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24814315 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24814334 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24814325 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05165097 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24814318 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05142046 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24814354 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05219162 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05219160 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24814336 |
- |
- |