| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600022217.mm.1 |
TC0600002898.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600022217.mm.1 |
NR_045986.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022217.mm.1 |
NR_045986.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022217.mm.1 |
NR_045987.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022217.mm.1 |
NR_045987.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022215.mm.1 |
TC0600002898.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600022215.mm.1 |
NR_045986.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022215.mm.1 |
NR_045986.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022215.mm.1 |
NR_045987.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022215.mm.1 |
NR_045987.1 |
Refseq |