Search
Search

Gm9871| Gene Symbol: Gm9871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027989.1 | 3 - 4 | 506 | 66 | ||
| GenBank mRNA | AK027905.1 | - | 3 - 4 | 506 | 66 | |
| BC125571.1 | - | 2 - 3 | 139 | 66 | ||
| BC144923.1 | - | 257 | 66 | |||
| BC145947.1 | - | 2 - 3 | 139 | 66 | ||
Set Membership: |
|