Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01190997_m1
          See other SLC4A5 GE Assays ›
          Gene Symbol:
          Slc4a5
          Gene Name:
          solute carrier family 4, sodium bicarbonate cotransporter, member 5
          Gene Aliases:
          C330006M16, C330016K18Rik
          Chromosome Location:
          Chr.6: 83207580 - 83304945 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01190997_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc4a5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001166067.1 NP_001159539.1 24 - 25 3500 63
          GenBank mRNA AK049140.1 - 7 - 8 789 63
          AK049248.1 - 11 - 12 1447 63
          BB398090.2 - 3 - 4 207 63
          CK626745.1 - 5 - 6 505 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000926.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Transporter 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          epithelial cell development
          renal system process
          regulation of systemic arterial blood pressure
          ion transport
          regulation of gene expression
          inorganic anion transport
          bicarbonate transport
          cerebrospinal fluid secretion
          sodium ion transmembrane transport
          mitochondrion distribution
          regulation of intracellular pH
          retina development in camera-type eye
          anion transmembrane transport
          molecular_function
          sodium:bicarbonate symporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032548 CK626745.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032548 AK049248.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032548 NM_001166067.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032548 AK049140.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403814 AK049248.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403814 BB398090.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403814 NM_001166067.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403814 AK049140.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403814 AK049248.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403814 BB398090.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403814 NM_001166067.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403814 AK049140.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106554 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179807 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123956 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403814 AK049248.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403814 BB398090.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403814 NM_001166067.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403814 AK049140.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403814 AK049248.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403814 BB398090.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403814 NM_001166067.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403814 AK049140.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032548 CK626745.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032548 AK049248.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032548 NM_001166067.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032548 AK049140.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106554 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05961241 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179807 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123956 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05961255 - -
          Whole Mouse Genome G4122A A_51_P403814 AK049248.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403814 BB398090.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403814 NM_001166067.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403814 AK049140.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403814 AK049248.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403814 BB398090.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403814 NM_001166067.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403814 AK049140.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P384831 AK049248.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P384831 CA526642.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P384831 CF725373.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P384831 NM_001166067.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106554 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123956 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05640030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05485453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05130865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05845263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05640031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05658906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05188823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05640035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05236169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05384889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05154139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05017656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05721595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05640033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05721597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05384887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05619795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05181360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24592532 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1435420_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10539393 - -
          Mouse Genome 430 2.0 Array 1435420_at - -
          Mouse Genome 430 2.0 Array 1435420_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600007083.mm.1 TC0600000926.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600007083.mm.1 NM_001166067.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007083.mm.1 NM_001166067.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007084.mm.1 TC0600000926.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600007084.mm.1 NM_001166067.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600007084.mm.1 NM_001166067.1 Refseq
          Mouse Expression Set 430 1435420_PM_at - -
          Mouse Expression Set 430 1435420_at - -
          Murine Genome U74v2 Set 96599_at - -
          Murine Genome U74v2 Set 96599_PM_at - -
          Mouse Genome 430A 2.0 Array 1435420_at - -
          Mouse Genome 430A 2.0 Array 1435420_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1435420_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1435420_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 617257 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2683820 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1249100 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2683820 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1216441 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline