Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01191246_g1
          See other E130218I03RIK GE Assays ›
          Gene Symbol:
          E130218I03Rik
          Gene Name:
          RIKEN cDNA E130218I03 gene
          Gene Aliases:
          E130106C14Rik
          Chromosome Location:
          Chr.4: 134243763 - 134245873 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01191246_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: E130218I03Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040436.1 4 - 5 504 73
          GenBank mRNA AK021371.1 - 4 - 5 505 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001630.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2195859 AK087464.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2195859 AK156931.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2195859 BC003465.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2195859 AK133585.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2195859 NM_024215.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 244503 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2195859 AK087464.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2195859 AK156931.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2195859 BC003465.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2195859 AK133585.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2195859 NM_024215.2 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1435318_PM_at - -
          Mouse Genome 430 2.0 Array 1435318_PM_at - -
          Mouse Genome 430 2.0 Array 1435318_at - -
          Affymetrix Mouse Clariom S Array PSR0400012956.mm.1 NR_040435.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012956.mm.1 NR_040435.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012956.mm.1 NR_040436.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012956.mm.1 NR_040436.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012956.mm.1 TC0400001630.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400012947.mm.1 NR_040435.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012947.mm.1 NR_040435.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012947.mm.1 NR_040436.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012947.mm.1 NR_040436.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012947.mm.1 TC0400001630.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400012942.mm.1 NR_040435.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012942.mm.1 NR_040435.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012942.mm.1 NR_040436.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012942.mm.1 NR_040436.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012942.mm.1 TC0400001630.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400012948.mm.1 NR_040435.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012948.mm.1 NR_040435.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012948.mm.1 NR_040436.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012948.mm.1 NR_040436.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012948.mm.1 TC0400001630.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400012949.mm.1 NR_040435.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012949.mm.1 NR_040435.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012949.mm.1 NR_040436.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012949.mm.1 NR_040436.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012949.mm.1 TC0400001630.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400012941.mm.1 NR_040436.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012941.mm.1 NR_040436.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012941.mm.1 TC0400001630.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400012954.mm.1 NR_040435.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012954.mm.1 NR_040435.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012954.mm.1 NR_040436.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012954.mm.1 NR_040436.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012954.mm.1 TC0400001630.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400012955.mm.1 NR_040435.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012955.mm.1 NR_040435.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012955.mm.1 NR_040436.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012955.mm.1 NR_040436.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012955.mm.1 TC0400001630.mm.1 MTA TC
          Mouse Expression Set 430 1435318_at - -
          Mouse Expression Set 430 1435318_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435318_at - -
          Affymetrix HT Mouse Genome 430B Array 1435318_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 635226 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline