| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000000526.mm.1 |
NR_046010.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000000526.mm.1 |
NR_046010.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000000526.mm.1 |
TC1000000078.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000000524.mm.1 |
NR_046010.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000000524.mm.1 |
NR_046010.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000000524.mm.1 |
TC1000000078.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000000525.mm.1 |
NR_046010.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000000525.mm.1 |
NR_046010.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000000525.mm.1 |
TC1000000078.mm.1 |
MTA TC |