Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01191815_m1
          See other LURAP1 GE Assays ›
          Gene Symbol:
          Lurap1
          Gene Name:
          leucine rich adaptor protein 1
          Gene Aliases:
          1520402A15Rik, Lrp35a
          Chromosome Location:
          Chr.4: 116136668 - 116144616 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01191815_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lurap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026547.1 NP_080823.1 1 - 2 318 131
          GenBank mRNA AK013564.1 - 1 - 2 318 131
          BC147058.1 - 1 - 2 275 9056069 131
          BC147059.1 - 1 - 2 275 9056037 131
          BC147737.1 - 1 - 2 275 9056686 131
          BC147750.1 - 1 - 2 275 9056699 131

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003333.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cytokine production
          cell migration
          actomyosin structure organization
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P547491 BC147059.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P547491 BC147737.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P547491 BC147750.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P547491 NM_026547.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P547491 AK013564.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P547491 BC147059.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P547491 BC147737.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P547491 BC147750.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P547491 NM_026547.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P547491 AK013564.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 342979 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 717332 AF193810.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 717332 AK076757.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 717332 NM_019799.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 671433 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102280 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P547491 BC147059.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P547491 BC147737.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P547491 BC147750.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P547491 NM_026547.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P547491 AK013564.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P547491 BC147059.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P547491 BC147737.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P547491 BC147750.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P547491 NM_026547.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P547491 AK013564.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P547491 BC147059.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P547491 BC147737.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P547491 BC147750.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P547491 NM_026547.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P547491 AK013564.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P547491 BC147059.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P547491 BC147737.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P547491 BC147750.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P547491 NM_026547.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P547491 AK013564.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P03978846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23227915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03399022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23227919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03978844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03978839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03978837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23227921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03978850 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10515279 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515278 - -
          Affymetrix HT MG-430 PM Array Plate 1430613_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10515277 - -
          Mouse Genome 430 2.0 Array 1430613_at - -
          Mouse Genome 430 2.0 Array 1430613_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68075_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68075_at - -
          Affymetrix Mouse Clariom S Array PSR0400026586.mm.1 TC0400003333.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400026586.mm.1 NM_026547.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026586.mm.1 NM_026547.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026583.mm.1 TC0400003333.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400026583.mm.1 NM_026547.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026583.mm.1 NM_026547.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026584.mm.1 TC0400003333.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400026584.mm.1 NM_026547.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026584.mm.1 NM_026547.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026585.mm.1 TC0400003333.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400026585.mm.1 NM_026547.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026585.mm.1 NM_026547.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026587.mm.1 TC0400003333.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400026587.mm.1 NM_026547.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026587.mm.1 NM_026547.1 Refseq
          Mouse Expression Set 430 1430613_at - -
          Mouse Expression Set 430 1430613_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430613_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430613_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 837625 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3850408 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248601 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1241455 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248601 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0