Search
Search

Ppil4| Gene Symbol: Ppil4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026141.3 | NP_080417.2 | 8 - 9 | 866 | 91 | |
| GenBank mRNA | AK011443.1 | - | 8 - 9 | 838 | 91 | |
| AK012371.1 | - | 8 - 9 | 843 | 91 | ||
| AK017370.1 | - | 8 - 9 | 866 | 91 | ||
| AK167168.1 | - | 8 - 9 | 831 | 91 | ||
| AK167258.1 | - | 8 - 9 | 838 | 91 | ||
| BC079912.1 | - | 8 - 9 | 828 | 30661438 | 91 | |
Set Membership: |
|