Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01192717_m1
          See other ZBTB8A GE Assays ›
          Gene Symbol:
          Zbtb8a
          Gene Name:
          zinc finger and BTB domain containing 8a
          Gene Aliases:
          2410081M15Rik, AI480941
          Chromosome Location:
          Chr.4: 129353632 - 129378028 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01192717_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zbtb8a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028603.4 NP_082879.1 4 - 5 1362 66
          GenBank mRNA AK010736.1 - 4 - 5 1361 66
          BC017614.1 - 4 - 5 1193 4912757 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003593.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zbtb8a

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          biological_process
          nucleic acid binding
          DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23296401 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23296400 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23296398 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23296399 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23296405 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23296403 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23296404 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P435817 BC017614.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P435817 NM_028603.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P435817 AK010736.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P435817 BC017614.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P435817 NM_028603.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P435817 AK010736.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 196893 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930711 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 937419 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 830915 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 277941 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 154495 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 958284 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 114121 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774060 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 13834 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431596 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358977 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 182169 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 557490 AK018320.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 692741 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35968 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 67901 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P132154 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116687 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P435817 BC017614.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P435817 NM_028603.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P435817 AK010736.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P435817 BC017614.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P435817 NM_028603.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P435817 AK010736.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296397 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296406 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296405 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296410 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00986715 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P132154 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00986723 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116687 - -
          Whole Mouse Genome G4122A A_51_P435817 BC017614.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P435817 NM_028603.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P435817 AK010736.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P435817 BC017614.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P435817 NM_028603.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P435817 AK010736.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115354 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P132154 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116687 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03687878 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03687870 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03687872 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03687871 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03687877 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23296401 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23296405 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23296399 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23296400 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23296403 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23296398 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23296404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04093674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296413 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03731934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03755608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03942347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03942348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03662220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03726218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03252267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296430 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03687871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296416 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03942352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03706243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04005671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296403 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03727191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296423 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10516593 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516591 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516592 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516595 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516594 - -
          Affymetrix HT MG-430 PM Array Plate 1452063_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10516590 - -
          Mouse Genome 430 2.0 Array 1452063_at - -
          Mouse Genome 430 2.0 Array 1452063_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73680_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73680_at - -
          Affymetrix Mouse Clariom S Array PSR0400029285.mm.1 TC0400003593.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400029285.mm.1 NM_028603.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029285.mm.1 NM_028603.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029284.mm.1 TC0400003593.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400029284.mm.1 NM_028603.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029284.mm.1 NM_028603.4 Refseq
          Mouse Expression Set 430 1452063_at - -
          Mouse Expression Set 430 1452063_PM_at - -
          Murine Genome U74v2 Set 135180_PM_at - -
          Murine Genome U74v2 Set 135180_at - -
          Mouse Genome 430A 2.0 Array 1452063_PM_at - -
          Mouse Genome 430A 2.0 Array 1452063_at - -
          Affymetrix HT Mouse Genome 430A Array 1452063_at - -
          Affymetrix HT Mouse Genome 430A Array 1452063_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 760220 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4280100 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004280100 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2753530 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2753530 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240747 - -
          mouse-6 v1.1 (Illumina) scl23783.5.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 4280100 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23783.5.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4280100 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2854121 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2854117 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2854121 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2854117 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline