Search
Search

Psme4| Gene Symbol: Psme4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134013.3 | NP_598774.2 | 44 - 45 | 5472 | 64 | |
| GenBank mRNA | AK129049.1 | - | 3 - 4 | 3451 | 64 | |
| AK169447.1 | - | 3 - 4 | 369 | 64 | ||
| BC004575.1 | - | 2 - 3 | 301 | 64 | ||
| BC024484.1 | - | 5 - 6 | 569 | 64 | ||
| BC031174.1 | - | 16 - 17 | 1916 | 64 | ||
| BC054364.1 | - | 2 - 3 | 218 | 64 | ||
| BC071256.1 | - | 15 - 16 | 1804 | 64 | ||
| BC141381.1 | - | 44 - 45 | 5185 | 64 | ||
Set Membership: |
|