Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Asb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023906.3 | NP_076395.2 | 7 - 8 | 1169 | 102 | |
GenBank mRNA | AF155354.1 | - | 6 - 7 | 984 | 102 | |
AK010315.1 | - | 7 - 8 | 1105 | 102 | ||
AK038086.1 | - | 7 - 8 | 1150 | 102 | ||
AK143580.1 | - | 7 - 8 | 1104 | 102 | ||
BC023086.1 | - | 7 - 8 | 1103 | 4947610 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |