Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01193518_m1
          See other SMIM23 GE Assays ›
          Gene Symbol:
          Smim23
          Gene Name:
          small integral membrane protein 23
          Gene Aliases:
          1700008A04Rik
          Chromosome Location:
          Chr.11: 32820376 - 32824594 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01193518_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smim23
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027050.1 NP_081326.1 3 - 4 291 65
          GenBank mRNA AK005748.1 - 3 - 4 271 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002480.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2075569 NM_027050.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2075569 AK005748.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2075569 EU477371.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P204898 NM_027050.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P204898 AK005748.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P204898 NM_027050.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P204898 AK005748.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148290 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2075569 NM_027050.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2075569 AK005748.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2075569 EU477371.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P204898 NM_027050.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P204898 AK005748.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P204898 NM_027050.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P204898 AK005748.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148290 - -
          Whole Mouse Genome G4122A A_51_P204898 NM_027050.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P204898 AK005748.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P204898 NM_027050.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P204898 AK005748.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09498389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09294289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09226574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09294294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09294297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08977446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09376429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09636447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08989978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09498390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09010028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09666967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09592635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08989980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09294292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09592642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09139746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09010667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09668150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27691144 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1429989_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10385027 - -
          Mouse Genome 430 2.0 Array 1429989_at - -
          Mouse Genome 430 2.0 Array 1429989_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69351_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69351_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100023100.mm.1 NM_027050.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023100.mm.1 NM_027050.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023100.mm.1 TC1100002480.mm.1 MTA TC
          Mouse Expression Set 430 1429989_at - -
          Mouse Expression Set 430 1429989_PM_at - -
          Murine Genome U74v2 Set 168879_PM_i_at - -
          Murine Genome U74v2 Set 168879_i_at - -
          Affymetrix HT Mouse Genome 430B Array 1429989_at - -
          Affymetrix HT Mouse Genome 430B Array 1429989_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 459553 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6220093 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2694516 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2694516 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline