Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10368692 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10368694 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10368691 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10368693 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10368690 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1444050_PM_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10368689 |
- |
- |
Mouse Genome 430 2.0 Array |
1444050_at |
- |
- |
Mouse Genome 430 2.0 Array |
1444050_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1000014324.mm.1 |
NM_001127376.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014324.mm.1 |
NM_001127376.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014324.mm.1 |
TC1000002018.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000014322.mm.1 |
NM_001127376.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014322.mm.1 |
NM_001127376.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014322.mm.1 |
TC1000002018.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000014323.mm.1 |
NM_001127376.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014323.mm.1 |
NM_001127376.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014323.mm.1 |
TC1000002018.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1444050_at |
- |
- |
Mouse Expression Set 430 |
1444050_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1444050_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1444050_PM_at |
- |
- |