Search
Search

Cyp4b1| Gene Symbol: Cyp4b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007823.2 | NP_031849.1 | 8 - 9 | 1092 | 83 | |
| GenBank mRNA | AK143890.1 | - | 8 - 9 | 1105 | 83 | |
| AK165445.1 | - | 8 - 9 | 1108 | 83 | ||
| BC008996.1 | - | 8 - 9 | 1096 | 3967680 | 83 | |
| D50834.1 | - | 8 - 9 | 1092 | 83 | ||
Set Membership: |
|