Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01193966_m1
          See other OSM GE Assays ›
          Gene Symbol:
          Osm
          Gene Name:
          oncostatin M
          Gene Aliases:
          OncoM
          Chromosome Location:
          Chr.11: 4236785 - 4241026 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01193966_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Osm
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013365.2 NP_001013383.1 2 - 3 266 82
          GenBank mRNA AK155637.1 - 2 - 3 217 82
          BC099866.1 - 2 - 3 218 40040107 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000040.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Osm

          Panther Classification:

          Molecular Function -

          cytokine interleukin superfamily intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          immune response
          peripheral nervous system development
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          response to heat
          positive regulation of peptidyl-serine phosphorylation
          oncostatin-M-mediated signaling pathway
          regulation of growth
          tyrosine phosphorylation of Stat3 protein
          tyrosine phosphorylation of Stat5 protein
          tyrosine phosphorylation of Stat1 protein
          positive regulation of tyrosine phosphorylation of Stat3 protein
          positive regulation of MAPK cascade
          negative regulation of meiotic nuclear division
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of hormone secretion
          behavioral response to pain
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of apoptotic signaling pathway
          cytokine activity
          oncostatin-M receptor binding
          growth factor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P291361 AK087945.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P291361 AK155637.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P291361 NM_001013365.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P291361 AK087945.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P291361 AK155637.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P291361 NM_001013365.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 577350 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43722 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 171331 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 127984 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166622 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P291361 AK087945.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P291361 AK155637.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P291361 NM_001013365.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P291361 AK087945.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P291361 AK155637.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P291361 NM_001013365.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P348614 AK155637.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P348614 BC099866.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P348614 NM_001013365.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P291361 AK087945.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P291361 AK155637.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P291361 NM_001013365.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P291361 AK087945.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P291361 AK155637.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P291361 NM_001013365.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27556543 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166622 - -
          Whole Mouse Genome G4122A A_52_P348614 AK155637.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P348614 BC099866.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P348614 NM_001013365.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P291361 AK087945.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P291361 AK155637.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P291361 NM_001013365.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P291361 AK087945.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P291361 AK155637.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P291361 NM_001013365.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166622 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09588061 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09138054 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09588062 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09554833 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09554828 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09138049 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09138055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27556557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09169582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09190039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09403707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09178148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09588052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09536963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09543630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09688978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09204256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09138055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27556522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09588057 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10373913 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373917 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373915 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373914 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373916 - -
          Affymetrix HT MG-430 PM Array Plate 1438767_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10373912 - -
          Mouse Genome 430 2.0 Array 1438767_at - -
          Mouse Genome 430 2.0 Array 1438767_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18413_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18413_at - -
          Affymetrix Mouse Clariom S Array PSR1100000512.mm.1 NM_001013365.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000512.mm.1 NM_001013365.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000512.mm.1 TC1100000040.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000513.mm.1 NM_001013365.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000513.mm.1 NM_001013365.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000513.mm.1 TC1100000040.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000514.mm.1 NM_001013365.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000514.mm.1 NM_001013365.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000514.mm.1 TC1100000040.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000515.mm.1 NM_001013365.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000515.mm.1 NM_001013365.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000515.mm.1 TC1100000040.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000516.mm.1 NM_001013365.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000516.mm.1 NM_001013365.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000516.mm.1 TC1100000040.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000517.mm.1 NM_001013365.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000517.mm.1 NM_001013365.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000517.mm.1 TC1100000040.mm.1 MTA TC
          Mouse Expression Set 430 1438767_at - -
          Mouse Expression Set 430 1438767_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1438767_at - -
          Affymetrix HT Mouse Genome 430B Array 1438767_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 563182 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106510484 - -
          MouseWG-6 v2 Expression BeadChip 100630577 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226916 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252325 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2543107 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2919411 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2919411 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0