Search
Search
| Gene Symbol: Wnt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009523.2 | NP_033549.1 | 1 - 2 | 123 | 68 | |
| GenBank mRNA | AK012727.1 | - | 1 - 2 | 250 | 68 | |
| BC101962.1 | - | 1 - 2 | 123 | 40044945 | 68 | |
| BC103560.1 | - | 1 - 2 | 123 | 40044943 | 68 | |
| BC103561.1 | - | 1 - 2 | 123 | 40044944 | 68 | |
| BC103562.1 | - | 1 - 2 | 123 | 40044946 | 68 | |
| M89797.1 | - | 1 - 2 | 123 | 68 | ||
Set Membership: |
|