Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01194005_g1
          See other CDC42 GE Assays ›
          Gene Symbol:
          Cdc42
          Gene Name:
          cell division cycle 42
          Gene Aliases:
          AI747189, AU018915
          Chromosome Location:
          Chr.4: 137319696 - 137357759 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01194005_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdc42
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001243769.1 NP_001230698.1 2 - 3 262 116
          NM_009861.3 NP_033991.1 2 - 3 262 116
          GenBank mRNA AK003098.1 - 2 - 3 183 116
          AK075567.1 - 2 - 3 181 116
          AK144216.1 - 2 - 3 185 116
          AK150563.1 - 2 - 3 196 116
          AK151087.1 - 2 - 3 207 116
          AK151726.1 - 2 - 3 184 116
          AK153564.1 - 2 - 3 206 116
          AK154870.1 - 2 - 3 181 116
          AK159470.1 - 2 - 3 224 116
          AK166281.1 - 2 - 3 181 116
          AK167195.1 - 2 - 3 170 116
          AK167400.1 - 2 - 3 175 116
          AK167609.1 - 2 - 3 180 116
          AK168013.1 - 1 - 2 175 116
          AK168076.1 - 2 - 3 175 116
          AK168089.1 - 1 - 2 175 116
          AK168276.1 - 2 - 3 175 116
          AK168503.1 - 2 - 3 196 116
          AK168758.1 - 2 - 3 215 116
          AK168820.1 - 2 - 3 188 116
          AK169122.1 - 2 - 3 196 116
          AK169232.1 - 2 - 3 175 116
          AK169805.1 - 2 - 3 200 116
          BC064792.1 - 2 - 3 168 30105331 116
          CD551112.1 - 2 - 3 249 116
          L11318.1 - 1 - 2 98 116
          L78075.1 - 1 - 2 102 116
          U37720.1 - 2 - 3 170 116

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdc42

          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          sprouting angiogenesis
          cardiac conduction system development
          keratinocyte development
          endocytosis
          actin filament organization
          Golgi organization
          regulation of mitotic nuclear division
          nuclear migration
          establishment or maintenance of cell polarity
          small GTPase mediated signal transduction
          Rho protein signal transduction
          nervous system development
          positive regulation of gene expression
          negative regulation of gene expression
          endosomal transport
          single organismal cell-cell adhesion
          cell differentiation
          hair follicle morphogenesis
          keratinization
          regulation of protein stability
          positive regulation of peptidyl-serine phosphorylation
          adherens junction organization
          cellular protein localization
          establishment or maintenance of apical/basal cell polarity
          multicellular organism growth
          dendritic cell migration
          regulation of protein catabolic process
          positive regulation of catalytic activity
          positive regulation of MAPK cascade
          regulation of protein heterodimerization activity
          positive regulation of neuron apoptotic process
          positive regulation of phosphatidylinositol 3-kinase activity
          positive regulation of DNA replication
          regulation of protein kinase activity
          positive regulation of JNK cascade
          filopodium assembly
          neuron fate determination
          epidermis morphogenesis
          actin filament bundle assembly
          regulation of protein metabolic process
          nucleus localization
          establishment of Golgi localization
          positive regulation of synapse structural plasticity
          heart contraction
          canonical Wnt signaling pathway
          epithelial-mesenchymal cell signaling
          hair follicle placode formation
          positive regulation of hair follicle cell proliferation
          organelle transport along microtubule
          actin filament branching
          epithelial cell-cell adhesion
          positive regulation of intracellular protein transport
          nucleotide binding
          GTPase activity
          protein binding
          GTP binding
          protein kinase binding
          GTP-dependent protein binding
          mitogen-activated protein kinase kinase kinase binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400031383.mm.1 TC0400003803.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400031383.mm.1 NM_001243769.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031383.mm.1 NM_001243769.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031383.mm.1 NM_009861.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031383.mm.1 NM_009861.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031389.mm.1 TC0400003803.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400031389.mm.1 NM_001243769.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031389.mm.1 NM_001243769.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031389.mm.1 NM_009861.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031389.mm.1 NM_009861.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031382.mm.1 TC0400003803.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400031382.mm.1 NM_001243769.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031382.mm.1 NM_001243769.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031382.mm.1 NM_009861.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031382.mm.1 NM_009861.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031385.mm.1 TC0400003803.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400031385.mm.1 NM_001243769.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031385.mm.1 NM_001243769.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031385.mm.1 NM_009861.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031385.mm.1 NM_009861.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031387.mm.1 TC0400003803.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400031387.mm.1 NM_001243769.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031387.mm.1 NM_001243769.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031387.mm.1 NM_009861.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031387.mm.1 NM_009861.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031380.mm.1 TC0400003803.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400031380.mm.1 NM_001243769.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031380.mm.1 NM_001243769.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031391.mm.1 TC0400003803.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400031391.mm.1 NM_001243769.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031391.mm.1 NM_001243769.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031391.mm.1 NM_009861.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031391.mm.1 NM_009861.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 486965 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline