Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01194064_m1
          See other SYNGR4 GE Assays ›
          Gene Symbol:
          Syngr4
          Gene Name:
          synaptogyrin 4
          Gene Aliases:
          1700016O14Rik
          Chromosome Location:
          Chr.7: 45886078 - 45896728 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01194064_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Syngr4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001291064.1 NP_001277993.1 2 - 3 299 98
          NM_021482.2 NP_067457.2 2 - 3 299 98
          GenBank mRNA AA231032.1 - 2 - 3 222 98
          AJ011734.1 - 98
          AK006050.1 - 2 - 3 254 98
          BC048469.1 - 2 - 3 299 6772653 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003027.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Syngr4

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2179805 BC048469.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2179805 AK006050.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2179805 NM_021482.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2179805 AJ011734.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P512627 BC048469.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P512627 BX635824.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P512627 AK006050.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P512627 NM_021482.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P512627 AJ011734.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P512627 BC048469.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P512627 BX635824.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P512627 AK006050.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P512627 NM_021482.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P512627 AJ011734.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 141148 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 502719 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 5985 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 686261 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 261114 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 40444 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 265008 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 126066 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160509 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2179805 BC048469.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2179805 AK006050.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2179805 NM_021482.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2179805 AJ011734.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P512627 BC048469.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P512627 BX635824.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P512627 AK006050.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P512627 NM_021482.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P512627 AJ011734.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P512627 BC048469.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P512627 BX635824.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P512627 AK006050.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P512627 NM_021482.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P512627 AJ011734.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25095702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25095721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25095706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25095716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25095724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25095700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25095722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25095712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25095704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25095717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25095701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25095709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25095714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25095726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25095711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25095718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25095699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25095695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25095719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25095696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25095694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25095707 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160509 - -
          Whole Mouse Genome G4122A A_51_P512627 BC048469.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512627 BX635824.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512627 AK006050.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512627 NM_021482.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512627 AJ011734.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512627 BC048469.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512627 BX635824.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512627 AK006050.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512627 NM_021482.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512627 AJ011734.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127645 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160509 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05987155 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06558842 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06558848 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06558835 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06558840 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06558845 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06558834 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06558846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25095712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06558848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06558846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06450765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06558839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06558837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25095712 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10563439 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563438 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563440 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563437 - -
          Affymetrix HT MG-430 PM Array Plate 1418875_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10563436 - -
          Mouse Genome 430 2.0 Array 1418875_at - -
          Mouse Genome 430 2.0 Array 1418875_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 58867_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 58867_at - -
          Affymetrix Mouse Clariom S Array PSR0700027510.mm.1 NM_021482.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027510.mm.1 NM_021482.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027510.mm.1 TC0700003027.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700027512.mm.1 NM_021482.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027512.mm.1 NM_021482.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027512.mm.1 TC0700003027.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700027506.mm.1 NM_021482.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027506.mm.1 NM_021482.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027506.mm.1 TC0700003027.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700027509.mm.1 NM_021482.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027509.mm.1 NM_021482.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027509.mm.1 TC0700003027.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700027507.mm.1 NM_021482.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027507.mm.1 NM_021482.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027507.mm.1 TC0700003027.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700027508.mm.1 NM_021482.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027508.mm.1 NM_021482.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027508.mm.1 TC0700003027.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700027513.mm.1 NM_021482.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027513.mm.1 NM_021482.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027513.mm.1 TC0700003027.mm.1 MTA TC
          Mouse Expression Set 430 1418875_at - -
          Mouse Expression Set 430 1418875_PM_at - -
          Murine Genome U74v2 Set 99531_PM_at - -
          Murine Genome U74v2 Set 99531_at - -
          Mouse Genome 430A 2.0 Array 1418875_PM_at - -
          Mouse Genome 430A 2.0 Array 1418875_at - -
          Affymetrix HT Mouse Genome 430A Array 1418875_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418875_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 744489 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6520450 - -
          Illumina mouse-6 v1.0 expression beadchip 102680687 - -
          Illumina mouse-6 v1.0 expression beadchip 6520450 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006520450 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1223537 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2766123 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223537 - -
          mouse-6 v1.1 (Illumina) scl00051.1_17-S - -
          mouse-6 v1.1 (Illumina) scl31357.5.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 102680687 - -
          Illumina Sentrix mouse-6.v1 bead array 6520450 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31357.5.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102680687 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6520450 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3148912 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1223537 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3070294 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3148912 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3070294 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1223537 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2766123 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline