Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01194236_m1
          See other SFRP5 GE Assays ›
          Gene Symbol:
          Sfrp5
          Gene Name:
          secreted frizzled-related sequence protein 5
          Gene Aliases:
          AI605071, SARP3
          Chromosome Location:
          Chr.19: 42197971 - 42202252 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01194236_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sfrp5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_018780.3 NP_061250.2 2 - 3 842 76
          GenBank mRNA BC032921.1 - 2 - 3 721 1395864 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001522.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sfrp5

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          negative regulation of cell proliferation
          Wnt signaling pathway
          cell differentiation
          negative regulation of Wnt signaling pathway
          negative regulation of catenin import into nucleus
          post-anal tail morphogenesis
          negative regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of JUN kinase activity
          digestive tract morphogenesis
          negative regulation of protein kinase B signaling
          convergent extension involved in axis elongation
          canonical Wnt signaling pathway
          negative regulation of canonical Wnt signaling pathway
          regulation of establishment of planar polarity
          planar cell polarity pathway involved in neural tube closure
          negative regulation of planar cell polarity pathway involved in axis elongation
          negative regulation of Wnt signaling pathway involved in digestive tract morphogenesis
          Wnt-protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119644 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119647 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119646 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119640 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119645 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119639 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119641 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119643 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2020203 BC032921.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 969296 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22080 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125942 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2020203 BC032921.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P476169 AF117759.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32119649 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125942 - -
          Whole Mouse Genome G4122A A_51_P476169 AF117759.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115373 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125942 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14355232 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14355208 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14355256 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119647 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119640 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119643 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119639 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119646 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119641 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119645 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32119644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32119635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14424554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14424565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14202886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14460068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14451889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14355238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14424549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14424544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14276314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14202887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14355217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14355223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14460066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14276306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14502045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14252587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14202882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14203145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14424570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14355256 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14282976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32119617 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10467745 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467748 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467747 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467746 - -
          Affymetrix HT MG-430 PM Array Plate 1436075_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1423023_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10467744 - -
          Mouse Genome 430 2.0 Array 1423023_PM_at - -
          Mouse Genome 430 2.0 Array 1423023_at - -
          Mouse Genome 430 2.0 Array 1436075_at - -
          Mouse Genome 430 2.0 Array 1436075_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 54612_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 54612_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900013429.mm.1 NM_018780.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013429.mm.1 NM_018780.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900013429.mm.1 TC1900001522.mm.1 MTA TC
          Mouse Expression Set 430 1423023_PM_at - -
          Mouse Expression Set 430 1423023_at - -
          Mouse Expression Set 430 1436075_at - -
          Mouse Expression Set 430 1436075_PM_at - -
          Mouse Genome 430A 2.0 Array 1423023_at - -
          Mouse Genome 430A 2.0 Array 1423023_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423023_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423023_at - -
          Affymetrix HT Mouse Genome 430B Array 1436075_at - -
          Affymetrix HT Mouse Genome 430B Array 1436075_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 717215 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2810114 - -
          Illumina mouse-6 v1.0 expression beadchip 2810114 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002810114 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232779 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232779 - -
          mouse-6 v1.1 (Illumina) scl52476.3.1_69-S - -
          Illumina Sentrix mouse-6.v1 bead array 2810114 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52476.3.1_69-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2810114 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1232779 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232779 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline