Search
Search

Fam189a2| Gene Symbol: Fam189a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114174.1 | NP_001107646.1 | 2 - 3 | 535 | 59 | |
| GenBank mRNA | BE994985.1 | - | 2 - 3 | 328 | 59 | |
Set Membership: |
|