Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01194496_m1
          See other IL31 GE Assays ›
          Gene Symbol:
          Il31
          Gene Name:
          interleukin 31
          Gene Aliases:
          1700013B14Rik
          Chromosome Location:
          Chr.5: 123480149 - 123482634 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01194496_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il31
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029594.1 NP_083870.1 2 - 3 219 123
          GenBank mRNA AK005939.1 - 2 - 3 389 123
          AY509149.1 - 2 - 3 219 123
          BC117082.1 - 2 - 3 218 40126401 123
          BC117084.1 - 2 - 3 218 40126403 123

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK005939.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          interleukin superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          cytokine activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P140397 NM_029594.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P140397 AK005939.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P140397 BC117082.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P140397 BC117084.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 581301 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131768 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137033 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163786 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P140397 NM_029594.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P140397 AK005939.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P140397 BC117082.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P140397 BC117084.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137033 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163786 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131768 - -
          Whole Mouse Genome G4122A A_52_P153179 NM_029594.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P153179 AK005939.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137033 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131768 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24055881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24055886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24055890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24055875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04493930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24055885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24055889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04135255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24055884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24055880 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24055882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24055877 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10533632 - -
          Affymetrix Mouse Gene 1.0 ST Array 10533629 - -
          Affymetrix Mouse Gene 1.0 ST Array 10533630 - -
          Affymetrix Mouse Gene 1.0 ST Array 10533631 - -
          Affymetrix HT MG-430 PM Array Plate 1430001_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10533628 - -
          Mouse Genome 430 2.0 Array 1430001_PM_at - -
          Mouse Genome 430 2.0 Array 1430001_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76399_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76399_at - -
          Affymetrix Mouse Clariom S Array PSR0500029036.mm.1 TC0500003233.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500029036.mm.1 NM_029594.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029036.mm.1 NM_029594.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029037.mm.1 TC0500003233.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500029037.mm.1 NM_029594.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029037.mm.1 NM_029594.1 Refseq
          Mouse Expression Set 430 1430001_at - -
          Mouse Expression Set 430 1430001_PM_at - -
          Murine Genome U74v2 Set 170153_r_at - -
          Murine Genome U74v2 Set 170153_PM_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1430001_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430001_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 779057 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1240632 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240632 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline