Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01195284_m1
          See other APOL8 GE Assays ›
          Gene Symbol:
          Apol8
          Gene Name:
          apolipoprotein L 8
          Gene Aliases:
          9830006J20Rik, APOL-II, Apol2
          Chromosome Location:
          Chr.15: 77745109 - 77757044 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01195284_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apol8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081970.1 NP_001075439.1 3 - 4 234 85
          GenBank mRNA AK036408.1 - 3 - 4 254 85
          AK046043.1 - 3 - 4 363 85
          BC129972.1 - 3 - 4 234 40090463 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001818.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Transporter 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          apolipoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 966359 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113023 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113023 - -
          Whole Mouse Genome G4122A A_52_P536096 AK046043.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P536096 BC129972.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P536096 NM_001081970.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P536096 AK036408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115471 AK046043.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115471 AK036408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115471 AK046043.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115471 AK036408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115471 AK046043.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115471 AK036408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115471 AK046043.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115471 AK036408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115471 AK046043.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115471 AK036408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115471 AK046043.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115471 AK036408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115471 AK046043.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115471 AK036408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115471 AK046043.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115471 AK036408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115471 AK046043.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115471 AK036408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115471 AK046043.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115471 AK036408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115471 AK046043.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115471 AK036408.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30355499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30355490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30355505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12377536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30355501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30355495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30355494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30355506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30355498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12311591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30355492 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10430198 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430196 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430199 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430197 - -
          Affymetrix HT MG-430 PM Array Plate 1441054_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10430195 - -
          Mouse Genome 430 2.0 Array 1441054_PM_at - -
          Mouse Genome 430 2.0 Array 1441054_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 239552_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 239552_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500014103.mm.1 NM_001081970.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014103.mm.1 NM_001081970.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014103.mm.1 TC1500001818.mm.1 MTA TC
          Mouse Expression Set 430 1441054_PM_at - -
          Mouse Expression Set 430 1441054_at - -
          Affymetrix HT Mouse Genome 430B Array 1441054_at - -
          Affymetrix HT Mouse Genome 430B Array 1441054_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 750050 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1260031 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1260031 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline