Search
Search

Zfp655| Gene Symbol: Zfp655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028298.3 | NP_082574.1 | 2 - 3 | 229 | 64 | |
| GenBank mRNA | AK012336.1 | - | 2 - 3 | 232 | 64 | |
| AK220552.1 | - | 2 - 3 | 234 | 64 | ||
| BC034731.1 | - | 2 - 3 | 232 | 64 | ||
| BC060660.1 | - | 2 - 3 | 265 | 30355779 | 64 | |
| BC062945.1 | - | 2 - 3 | 229 | 30357186 | 64 | |
Set Membership: |
|