Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01197335_m1
          See other KLK13 GE Assays ›
          Gene Symbol:
          Klk13
          Gene Name:
          kallikrein related-peptidase 13
          Gene Aliases:
          Egfbp-2, mGk-13
          Chromosome Location:
          Chr.7: 43712567 - 43727864 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01197335_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klk13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039042.2 NP_001034131.2 4 - 5 691 72
          GenBank mRNA BC063763.1 - 4 - 5 692 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000770.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Protease Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          protease serine protease protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          protein processing
          endopeptidase activity
          serine-type endopeptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2084965 BC063763.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2084965 NM_001039042.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 604364 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667823 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 272663 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 583765 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 261726 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 552891 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 957588 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 447619 AK006852.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 38413 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 8040 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159406 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147474 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2084965 BC063763.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2084965 NM_001039042.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083728 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06178184 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147474 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01505146 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159406 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159406 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06488423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06328809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06103876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06016716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06123950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06328805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06421638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06384059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06234700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06450289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06167438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06286419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06234699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06167441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083785 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552470 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552474 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552471 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552472 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552473 - -
          Affymetrix Mouse Gene 1.1 ST Array 10552469 - -
          Affymetrix Mouse Clariom S Array PSR0700006251.mm.1 TC0700000770.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006251.mm.1 NM_001039042.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006251.mm.1 NM_001039042.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006253.mm.1 TC0700000770.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006253.mm.1 NM_001039042.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006253.mm.1 NM_001039042.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006249.mm.1 TC0700000770.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006249.mm.1 NM_001039042.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006249.mm.1 NM_001039042.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006250.mm.1 TC0700000770.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006250.mm.1 NM_001039042.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006250.mm.1 NM_001039042.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006248.mm.1 TC0700000770.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006248.mm.1 NM_001039042.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006248.mm.1 NM_001039042.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006252.mm.1 TC0700000770.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006252.mm.1 NM_001039042.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006252.mm.1 NM_001039042.2 Refseq
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237292 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3115315 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3040588 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3040588 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3115315 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline