Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01197405_m1
          See other LRCOL1 GE Assays ›
          Gene Symbol:
          Lrcol1
          Gene Name:
          leucine rich colipase-like 1
          Gene Aliases:
          Clpsl3, Gm1679
          Chromosome Location:
          Chr.5: 110344507 - 110356094 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01197405_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrcol1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033459.3 NP_001028631.1 3 - 4 447 92
          GenBank mRNA AK141249.1 - 3 - 4 448 92
          BC147447.1 - 3 - 4 385 9056089 92
          BC147469.1 - 3 - 4 385 9056111 92
          BC171993.1 - 3 - 4 363 92
          JQ258939.1 - 5 - 6 592 92
          JQ258940.1 - 6 - 7 655 92
          JQ258941.1 - 5 - 6 852 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001142.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          digestion
          biological_process
          lipid catabolic process
          molecular_function
          enzyme activator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23979249 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23979248 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23979255 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23979251 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23979253 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23979250 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23979254 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23979252 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P110707 NM_001033459.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P110707 AK141249.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P110707 BC147447.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P110707 BC147469.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P110707 BC171993.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 226406 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P110707 NM_001033459.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P110707 AK141249.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P110707 BC147447.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P110707 BC147469.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P110707 BC171993.1 mouse_transcriptome
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23979255 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23979253 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23979251 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23979252 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23979254 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23979249 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23979250 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23979248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23979164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23979209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04301877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04138569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04197604 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23979177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23979202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04570873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04570872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23979189 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23979192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23979207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04665429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23979175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23979178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04211273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23979184 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23979167 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04556394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04628941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23979182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23979200 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23979208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23979191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23979176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04211272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23979213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23979190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04939517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23979217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04286323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04665420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23979215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04570869 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10524226 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524223 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524225 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524224 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524222 - -
          Affymetrix HT MG-430 PM Array Plate 1440419_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10524221 - -
          Mouse Genome 430 2.0 Array 1440419_at - -
          Mouse Genome 430 2.0 Array 1440419_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 381667_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 381667_at - -
          Affymetrix Mouse Clariom S Array PSR0500009814.mm.1 TC0500001142.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500009815.mm.1 TC0500001142.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500009816.mm.1 TC0500001142.mm.1 MTA TC
          Mouse Expression Set 430 1440419_PM_at - -
          Mouse Expression Set 430 1440419_at - -
          Affymetrix HT Mouse Genome 430B Array 1440419_at - -
          Affymetrix HT Mouse Genome 430B Array 1440419_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 548032 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225092 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2864485 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2864485 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline