Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01197916_m1
          See other WRN GE Assays ›
          Gene Symbol:
          Wrn
          Gene Name:
          Werner syndrome homolog (human)
          Gene Aliases:
          AI846146
          Chromosome Location:
          Chr.8: 33234372 - 33385545 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01197916_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wrn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001122822.1 NP_001116294.1 3 - 4 295 61
          NM_011721.4 NP_035851.3 3 - 4 405 61
          GenBank mRNA AF091215.1 - 3 - 4 359 61
          AF241636.1 - 3 - 4 420 61
          AK143778.1 - 3 - 4 383 61
          AK153824.1 - 4 - 5 488 61
          AK158180.1 - 3 - 4 266 61
          AK165679.1 - 3 - 4 652 61
          AK171490.1 - 3 - 4 406 61
          BC050921.1 - 3 - 4 350 6408901 61
          BC060700.1 - 3 - 4 350 6408901 61
          D86526.1 - 3 - 4 375 61
          D86527.1 - 3 - 4 375 61
          U97045.1 - 313 61

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the U97045.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Wrn

          Panther Classification:

          Molecular Function -

          DNA helicase

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance
          DNA synthesis involved in DNA repair
          replicative cell aging
          nucleobase-containing compound metabolic process
          DNA metabolic process
          DNA replication
          DNA repair
          base-excision repair
          double-strand break repair
          DNA recombination
          cellular response to DNA damage stimulus
          response to oxidative stress
          cell aging
          metabolic process
          cellular response to starvation
          response to UV-C
          multicellular organism aging
          replication fork processing
          nucleolus to nucleoplasm transport
          DNA duplex unwinding
          regulation of growth rate
          regulation of apoptotic process
          cellular metabolic process
          positive regulation of hydrolase activity
          cellular response to gamma radiation
          nucleic acid phosphodiester bond hydrolysis
          nucleotide binding
          magnesium ion binding
          Y-form DNA binding
          bubble DNA binding
          nucleic acid binding
          DNA binding
          DNA helicase activity
          catalytic activity
          ATP-dependent DNA helicase activity
          helicase activity
          nuclease activity
          exonuclease activity
          protein binding
          ATP binding
          ATP-dependent helicase activity
          3'-5' exonuclease activity
          four-way junction helicase activity
          hydrolase activity
          ATPase activity
          manganese ion binding
          protein complex binding
          phosphoric ester hydrolase activity
          protein homodimerization activity
          3'-5' DNA helicase activity
          ATP-dependent 3'-5' DNA helicase activity
          T/G mismatch-specific endonuclease activity
          retroviral 3' processing activity
          metal ion binding
          G-quadruplex DNA binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800015728.mm.1 NM_001122822.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800015728.mm.1 NM_001122822.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800015728.mm.1 NM_011721.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800015728.mm.1 NM_011721.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800015728.mm.1 TC0800002004.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 360112 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline