| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10448246 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10442289 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10442290 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10442293 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448244 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10448245 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10442291 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10442292 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10448243 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10442288 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1700002210.mm.1 |
NM_001104578.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002210.mm.1 |
NM_001104578.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002210.mm.1 |
TC1700000305.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002207.mm.1 |
NM_001104578.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002207.mm.1 |
NM_001104578.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002207.mm.1 |
TC1700000305.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002209.mm.1 |
NM_001104578.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002209.mm.1 |
NM_001104578.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002209.mm.1 |
TC1700000305.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002211.mm.1 |
NM_001104578.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002211.mm.1 |
NM_001104578.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002211.mm.1 |
TC1700000305.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002208.mm.1 |
NM_001104578.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002208.mm.1 |
NM_001104578.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002208.mm.1 |
TC1700000305.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002206.mm.1 |
NM_001104578.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002206.mm.1 |
NM_001104578.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002206.mm.1 |
TC1700000305.mm.1 |
MTA TC |