Search
Search
| Gene Symbol: Gak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282051.1 | NP_001268980.1 | 7 - 8 | 867 | 65 | |
| NM_001282052.1 | NP_001268981.1 | 6 - 7 | 729 | 65 | ||
| NM_153569.2 | NP_705797.1 | 7 - 8 | 872 | 65 | ||
| GenBank mRNA | AK051394.1 | - | 6 - 7 | 585 | 65 | |
| AK133581.1 | - | 6 - 7 | 708 | 65 | ||
| AK146695.1 | - | 7 - 8 | 836 | 65 | ||
| AK150112.1 | - | 5 - 6 | 676 | 65 | ||
| AK161151.1 | - | 6 - 7 | 691 | 65 | ||
| AK166682.1 | - | 7 - 8 | 851 | 65 | ||
| BC060622.1 | - | 7 - 8 | 825 | 6826081 | 65 | |
| BC062975.1 | - | 6 - 7 | 677 | 65 | ||
| BC065051.1 | - | 6 - 7 | 677 | 65 | ||
| BC065052.1 | - | 7 - 8 | 834 | 5705695 | 65 | |
| BC065109.1 | - | 7 - 8 | 835 | 65 | ||
Set Membership: |
|