Search
Search

Ccdc18| Gene Symbol: Ccdc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028481.1 | NP_082757.1 | 24 - 25 | 3428 | 74 | |
| GenBank mRNA | AK016519.1 | - | 24 - 25 | 3422 | 74 | |
| BC082598.1 | - | 24 - 25 | 3428 | 30641632 | 74 | |
Set Membership: |
|