Search
Search

Cwf19l1| Gene Symbol: Cwf19l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081077.1 | NP_001074546.1 | 9 - 10 | 1051 | 66 | |
| GenBank mRNA | AK088543.1 | - | 11 - 12 | 2293 | 66 | |
| AK139654.1 | - | 9 - 10 | 1052 | 66 | ||
| BC037640.1 | - | 9 - 10 | 1029 | 5096937 | 66 | |
Set Membership: |
|