Search
Search

Fndc7| Gene Symbol: Fndc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165965.1 | NP_001159437.1 | 10 - 11 | 2216 | 69 | |
| NM_177091.5 | NP_796065.2 | 11 - 12 | 2474 | 69 | ||
| GenBank mRNA | AK053989.1 | - | 11 - 12 | 2337 | 69 | |
| BC111898.1 | - | 10 - 11 | 2023 | 40061374 | 69 | |
| BC113188.1 | - | 11 - 12 | 2279 | 69 | ||
Set Membership: |
|